Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentafluorophenyl Trifluoroacetate

Catalog No. 3770745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-707-45G 5 g
37-707-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 37-707-45G Supplier Sigma Aldrich Supplier No. 3770745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentafluorophenyl trifluoroacetate can be used:In the esterification of 2'sa-carboxyrhodamine dye to form pentafluorophenyl ester as a single isomer.As an acylating agent and a coupling agent for the peptide-type coupling of N-substituted glycine oligomers.In the synthesis of pentafluorophenyl thiophene-3-acetate (PFPTA) by reacting with 3-thiopheneacetic acid.may contain a small amount of pentafluorophenol

Specifications

CAS 14533-84-7
Density 1.63 g/mL at 25°C
Boiling Point 122°C to 123°C
Molecular Formula C8F8O2
Refractive Index n20/D 1.368
Linear Formula CF3CO2C6F5
MDL Number MFCD00134438
Quantity 5 g
Synonym Trifluoroacetic acid pentafluorophenyl ester
Molecular Weight (g/mol) 280.07 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.