Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P19045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1904-5G 5 g
P19041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1904-5G Supplier TCI America Supplier No. P19045G
Add to Cart
Add to Cart

Specifications

CAS 1582-24-7
Melting Point 290°C
Color White
Molecular Formula C6H2BF5O2
Quantity 5 g
MDL Number MFCD01074663
Synonym 2,3,4,5,6-pentafluorobenzeneboronic acid, pentafluorophenylboronic acid, perfluorophenylboronic acid, pentafluorobenzeneboronic acid, boronic acid, pentafluorophenyl
InChI Key VASOMTXTRMYSKD-UHFFFAOYSA-N
SMILES B(C1=C(C(=C(C(=C1F)F)F)F)F)(O)O
IUPAC Name (2,3,4,5,6-pentafluorophenyl)boronic acid
Molecular Weight (g/mol) 211.882
PubChem CID 2775922
Formula Weight 211.88
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.