Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyldimethylsilyldiethylamine 98.0+%, TCI America™
SDP

Catalog No. P0908100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0908100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0908100MG Supplier TCI America Supplier No. P0908100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55485-74-0
Molecular Formula C12H16F5NSi
Molecular Weight (g/mol) 297.344
MDL Number MFCD00013548
InChI Key ZBVPABKIRWOTNJ-UHFFFAOYSA-N
Synonym Flophemesyldiethylamine
PubChem CID 2769343
IUPAC Name N-[dimethyl-(2,3,4,5,6-pentafluorophenyl)silyl]-N-ethylethanamine
SMILES CCN(CC)[Si](C)(C)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Quantity 100 mg
UN Number 3267
Formula Weight 297.34
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Pentafluorophenyldimethylsilyldiethylamine [Pentafluorophenyldimethylsilylating Agent]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.