Learn More
Strem, An Ascensus Company PDMAT min. 98% 1g
SDP
Supplier:  Strem, An Ascensus Company 7308001G
Encompass
CAS# 19824-59-0. 1g. Pentakis(dimethylamino)tantalum(V), min 98% PDMAT. MFCD01631286. Molecular Weight: 401.33. Molecular Formula: Ta[N(CH3)2]5. Color/form: orange xtl. Strem# 73-0800. http://www.strem.com/catalog/v/73-0800/
Catalog No. 50-901-12989