Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentamethoxy red | 1755-51-7 | MFCD00070616 | 98.0% | 410.46 g/mol | C24H26O6 | 250 MG
SDP

Catalog No. 5000282043
Encompass_Preferred

Pentamethoxy red is a biochemical assay reagent used in laboratory research for analytical assays and staining applications. It is supplied as a solid organic dye intended for research use only; handle and store the material under the conditions specified in the certificate of analysis.

  • Useful as a reagent for biochemical assays and staining
  • Molecular weight 410.46 g/mol
  • Chemical formula C24H26O6
  • CAS number 1755-51-7
  • Commonly supplied in 250 mg packages
  • Store under conditions recommended in the certificate of analysis

Catalog No. 50-002-82043 Supplier Medchemexpress LLC Supplier No. HYW273209250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentamethoxy red is a biochemical assay reagent used in laboratory research for analytical assays and staining applications. It is supplied as a solid organic dye intended for research use only; handle and store the material under the conditions specified in the certificate of analysis.

  • Useful as a reagent for biochemical assays and staining
  • Molecular weight 410.46 g/mol
  • Chemical formula C24H26O6
  • CAS number 1755-51-7
  • Commonly supplied in 250 mg packages
  • Store under conditions recommended in the certificate of analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.