Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentamethylbenzene, 99%

Catalog No. AAL0416306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0416306 5 g
AAL0416314 25 g
AAL0416322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0416306 Supplier Thermo Scientific Chemicals Supplier No. L0416306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 700-12-9 | C11H16 | 148.249 g/mol

Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentamethylbenzene is used to prepare a mixture of nitropentamethylbenzene and 2,3,4,5-tetramethylbenzyl nitrate1. It was also used to prepare propene.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-12-9
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008522
InChI Key BEZDDPMMPIDMGJ-UHFFFAOYSA-N
Synonym pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
PubChem CID 12784
ChEBI CHEBI:38998
IUPAC Name 1,2,3,4,5-pentamethylbenzene
SMILES CC1=CC(=C(C(=C1C)C)C)C

Specifications

Melting Point 50°C to 51°C
Density 0.917
Boiling Point 231°C to 232°C
Flash Point 91°C (195°F)
Quantity 5 g
UN Number UN1325
Beilstein 1905349
Formula Weight 148.25
Percent Purity 99%
Chemical Name or Material Pentamethylbenzene
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-837-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.