Learn More
Strem, An Ascensus Company CAS# 4045-44-7. 1g. Pentamethylcyclopentadiene, min. 98%. MFCD00001354
SDP
Catalog No. 5090108850
Encompass
CAS# 4045-44-7. 1g. Pentamethylcyclopentadiene, min. 98%. MFCD00001354. Molecular Weight: 136.24. Molecular Formula: C10H16. Color/form: pale yellow liq. Strem# 06-1290. http://www.strem.com/catalog/v/06-1290/
Catalog No. 50-901-08850 Supplier Strem, An Ascensus Company Supplier No. 0612901G
CAS# 4045-44-7. 1g. Pentamethylcyclopentadiene, min. 98%. MFCD00001354. Molecular Weight: 136.24. Molecular Formula: C10H16. Color/form: pale yellow liq. Strem# 06-1290. http://www.strem.com/catalog/v/06-1290/