Learn More
Strem, An Ascensus Company CAS# 75181-07-6. 1g. Pentamethylcyclopentadienylzirconium trichloride, 99%. MFCD00070444
SDP
Supplier:  Strem, An Ascensus Company 4015001G
Encompass
CAS# 75181-07-6. 1g. Pentamethylcyclopentadienylzirconium trichloride, 99%. MFCD00070444. Molecular Weight: 332.81. Molecular Formula: (CH3)5C5ZrCl3. Color/form: pale yellow xtl. Strem# 40-1500. http://www.strem.com/catalog/v/40-1500/
Catalog No. 50-901-11591