Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentamethylcyclopentadienyliridium(III) Chloride,Dimer

Catalog No. 357537500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-753-7500MG 500 mg
35-753-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-753-7500MG Supplier Sigma Aldrich Supplier No. 357537500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.Catalyst for greener amine synthesis from amines and alcohols by hydrogen-borrowing. Precursor to cooperative metal-Bronsted acid catalyst for greener reductive amination using hydrogen gas.Metal-Br++nsted Acid Cooperative Catalysis for Asymmetric Reductive AminationDevelopment of GSK's reagent guides - embedding sustainability into reagent selection

Specifications

CAS 12354-84-6
Molecular Formula C20H30Cl4Ir2
Linear Formula C20H30Cl4Ir2
MDL Number MFCD00075435
Quantity 500 mg
Molecular Weight (g/mol) 796.7 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.