Learn More
Cayman Chemical Pentamidine, Purity: ≥98%, CAS: 100-33-4, 100 mg
SDP
Catalog No. 502982341
Encompass_Preferred
An antimicrobial agent; binds A/T rich regions in DNA minor grooves; active against L. mexicana (IC50 = 4.532 µg/ml), the T. brucei strain S427 (IC50 = 1.1 nM), and C. neoformans (MICs = 1.56 and 6.25 µg/ml for the JEC21 and H99 strains, respectively); increases survival in mice infected with T. brucei when administered at a dose of 10 µmol/kg
Catalog No. 50-298-2341 Supplier Cayman Chemical Supplier No. 20679100 MG
An antimicrobial agent; binds A/T rich regions in DNA minor grooves; active against L. mexicana (IC50 = 4.532 µg/ml), the T. brucei strain S427 (IC50 = 1.1 nM), and C. neoformans (MICs = 1.56 and 6.25 µg/ml for the JEC21 and H99 strains, respectively); increases survival in mice infected with T. brucei when administered at a dose of 10 µmol/kg