Learn More
Medchemexpress LLC HY-B0537B 50mg , Pentamidine (isethionate) CAS:140-64-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0537B50MG
Encompass_Preferred
HY-B0537B 50mg Pentamidine (isethionate) CAS: 140-64-7 Pentamidine isethionate (MP-601205 isethionate) is an antimicrobial agent and interferes with DNA biosynthetics. Pentamidine isethionate inhibits parasite Leishmania infantum with an IC50 of 2.5 μM. Pentamidine isethionate is a potent and selective protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL) inhibitor. Pentamidine isethionate has the potential for Gambian trypanosomiasis, antimony-resistant leishmaniasis, and Pneumocystis carinii pneumonia treatment. Antitumor and antibacterial activities[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9814