Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentan-3-amine-d5 | 1219802-43-3 | 92.19 g/mol | C5H8D5N | 10 MG
SDP

Catalog No. 5000208317
Encompass_Preferred

Pentan-3-amine-d5 is the deuterium-labeled analogue of pentan-3-amine intended for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Deuteration can influence pharmacokinetic and metabolic profiles, making this compound useful in drug development and analytical workflows.

  • Use as a tracer for quantitation in analytical methods
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Available in multiple sizes: 5 mg, 10 mg, 50 mg, 100 mg
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Molecular formula C5H8D5N and molecular weight 92.19 g/mol
  • Appearance: colorless to light yellow liquid

Catalog No. 50-002-08317 Supplier Medchemexpress LLC Supplier No. HYW087960S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentan-3-amine-d5 is the deuterium-labeled analogue of pentan-3-amine intended for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Deuteration can influence pharmacokinetic and metabolic profiles, making this compound useful in drug development and analytical workflows.

  • Use as a tracer for quantitation in analytical methods
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Available in multiple sizes: 5 mg, 10 mg, 50 mg, 100 mg
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
  • Molecular formula C5H8D5N and molecular weight 92.19 g/mol
  • Appearance: colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.