Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentan-3-amine-d5 | 1219802-43-3 | 92.19 g/mol | C5H8D5N | 5 MG
SDP

Catalog No. 5000208367
Encompass_Preferred

Pentan-3-amine-d5 is the deuterium-labeled isotopologue of pentan-3-amine used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a liquid that is colorless to light yellow and is intended for analytical applications requiring isotopically labeled standards.

  • Deuterium-labeled pentan-3-amine for tracer and internal standard use.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular formula C5H8D5N and molecular weight 92.19 g/mol.
  • Physical form: liquid, colorless to light yellow.
  • Storage guidance includes -20°C for long-term stability; in solvent, store at -80°C for extended periods.
  • Available in a small research-scale quantity (5 mg).

Catalog No. 50-002-08367 Supplier Medchemexpress LLC Supplier No. HYW087960S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentan-3-amine-d5 is the deuterium-labeled isotopologue of pentan-3-amine used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is supplied as a liquid that is colorless to light yellow and is intended for analytical applications requiring isotopically labeled standards.

  • Deuterium-labeled pentan-3-amine for tracer and internal standard use.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular formula C5H8D5N and molecular weight 92.19 g/mol.
  • Physical form: liquid, colorless to light yellow.
  • Storage guidance includes -20°C for long-term stability; in solvent, store at -80°C for extended periods.
  • Available in a small research-scale quantity (5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.