Learn More
Cayman Chemical PentaNacetylchItopentaose 10mg
SDP
Catalog No. 502971166
Encompass_Preferred
A pentameric chito-oligosaccharide involved in root nodulation; produced by NodC, a chito-oligosaccharide synthase, and serves as a substrate for NodL, an O-acetyltransferase; binds plant root lectins, an interaction that may be important in the promotion of cell division in the root cortex
Catalog No. 50-297-1166 Supplier Cayman Chemical Supplier No. 1610110 MG
A pentameric chito-oligosaccharide involved in root nodulation; produced by NodC, a chito-oligosaccharide synthase, and serves as a substrate for NodL, an O-acetyltransferase; binds plant root lectins, an interaction that may be important in the promotion of cell division in the root cortex