Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentanamide, 2-amino-4-methyl-, hydrochloride (1:1), (2S)- | 10466-61-2 | MFCD00013012 | 97.0% | 166.65 g·mol⁻¹ | C6H15ClN2O | 25 G
SDP

Catalog No. 5000209705
Encompass_Preferred

L-Leucinamide hydrochloride is the hydrochloride salt of L-leucinamide, an amino acid derivative reported to have anti-inflammatory properties. The product is supplied as a solid for research use, with specified purity and recommended storage conditions.

  • Hydrochloride salt, solid form.
  • Purity 97.0% (NMR).
  • Molecular formula C6H15ClN2O, molecular weight 166.65 g·mol⁻¹.
  • CAS number 10466-61-2.
  • Packaging: 25 g; recommended storage: powder at -20°C for up to 3 years or 4°C for up to 2 years.

Catalog No. 50-002-09705 Supplier Medchemexpress LLC Supplier No. 25GHYW088014
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucinamide hydrochloride is the hydrochloride salt of L-leucinamide, an amino acid derivative reported to have anti-inflammatory properties. The product is supplied as a solid for research use, with specified purity and recommended storage conditions.

  • Hydrochloride salt, solid form.
  • Purity 97.0% (NMR).
  • Molecular formula C6H15ClN2O, molecular weight 166.65 g·mol⁻¹.
  • CAS number 10466-61-2.
  • Packaging: 25 g; recommended storage: powder at -20°C for up to 3 years or 4°C for up to 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.