Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentanamide, 2-amino-5-[(aminoiminomethyl)amino]-N-(4-nitrophenyl)-, dihydrochloride, (2S)- | 40127-11-5 | 99.8% | 367.23 | 100 MG
SDP

Catalog No. 5000351214
Encompass_Preferred

L-Arginine p-nitroanilide dihydrochloride is a chromogenic substrate specifically designed for cathepsin H and aminopeptidases. This product is provided as an off-white to light yellow solid.

  • Chromogenic substrate
  • Suitable for cathepsin H and aminopeptidases
  • Soluble in H2O at 200 mg/mL
  • Recommended storage at 4°C, sealed and away from moisture
  • For research use only

Catalog No. 50-003-51214 Supplier Medchemexpress LLC Supplier No. HY137895100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Arginine p-nitroanilide dihydrochloride is a chromogenic substrate specifically designed for cathepsin H and aminopeptidases. This product is provided as an off-white to light yellow solid.

  • Chromogenic substrate
  • Suitable for cathepsin H and aminopeptidases
  • Soluble in H2O at 200 mg/mL
  • Recommended storage at 4°C, sealed and away from moisture
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.