Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentane-1,3,5-triol | 4328-94-3 | MFCD15146836 | ≥97.0% | 120.15 g/mol | C5H12O3 | 100 MG
SDP

Catalog No. 5000285247
Encompass_Preferred

Pentane-1,3,5-triol is a trihydroxylated aliphatic compound supplied for research use as a chemical building block and drug intermediate in medicinal chemistry and organic synthesis workflows.

  • CAS number: 4328-94-3.
  • Chemical formula: C5H12O3.
  • Molecular weight: 120.15 g/mol.
  • Purity: ≥97.0%.
  • Available in small pack sizes suitable for laboratory synthesis (100 mg to 10 g).
  • Commonly used as a synthetic intermediate in the preparation of active compounds.

Catalog No. 50-002-85247 Supplier Medchemexpress LLC Supplier No. HYW358956100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentane-1,3,5-triol is a trihydroxylated aliphatic compound supplied for research use as a chemical building block and drug intermediate in medicinal chemistry and organic synthesis workflows.

  • CAS number: 4328-94-3.
  • Chemical formula: C5H12O3.
  • Molecular weight: 120.15 g/mol.
  • Purity: ≥97.0%.
  • Available in small pack sizes suitable for laboratory synthesis (100 mg to 10 g).
  • Commonly used as a synthetic intermediate in the preparation of active compounds.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.