Learn More
Medchemexpress LLC Pentane-1,5-diamine dihydrochloride | 1476-39-7 | 99.9% | 175.10 g·mol⁻¹ | C5H16Cl2N2 | 500 MG
SDP
Catalog No. 5000301154
Encompass_Preferred
Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of pentane-1,5-diamine, an endogenous diamine metabolite supplied as a high-purity solid for biochemical and pharmacological research. It is used in studies of polyamine metabolism and related biological pathways and is offered in both solid and DMSO solution formats.
  • High purity: 99.90%.
  • Well characterized: formula C5H16Cl2N2 and molecular weight 175.10 g·mol⁻¹.
  • Available as solid and as 10 mM solutions in DMSO.
  • Suitable for biochemical and pharmacological research applications.
  • Multiple pack sizes for flexibility in experimental design.
Catalog No. 50-003-01154 Supplier Medchemexpress LLC Supplier No. HYW016750500MG
Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of pentane-1,5-diamine, an endogenous diamine metabolite supplied as a high-purity solid for biochemical and pharmacological research. It is used in studies of polyamine metabolism and related biological pathways and is offered in both solid and DMSO solution formats.
  • High purity: 99.90%.
  • Well characterized: formula C5H16Cl2N2 and molecular weight 175.10 g·mol⁻¹.
  • Available as solid and as 10 mM solutions in DMSO.
  • Suitable for biochemical and pharmacological research applications.
  • Multiple pack sizes for flexibility in experimental design.