Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentane-1,5-diamine dihydrochloride | 1476-39-7 | MFCD00012527 | 98.0% | 175.10 g/mol | C5H16Cl2N2 | 1 ML
SDP

Catalog No. 5000301874
Encompass_Preferred

Pentane-1,5-diamine dihydrochloride is an endogenous diamine metabolite provided as research-grade reference material for analytical and synthetic laboratory use. It serves as an analytical standard in chromatographic and mass-spectrometry workflows and is offered in both solid and ready-to-use solution formats for flexibility in method development and QC applications.

  • Analytical standard suitable for HPLC, GC, and MS.
  • Available as a 10 mM solution in DMSO and in multiple solid pack sizes.
  • Molecular weight 175.10 g/mol; formula C5H16Cl2N2; CAS 1476-39-7.
  • Standard grade purity 98% for solid/reference material.
  • Documentation available: data sheet, COA, SDS, and spectral files.
  • Intended for research use only; follow handling and storage instructions.

Catalog No. 50-003-01874 Supplier Medchemexpress LLC Supplier No. HYW01675010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentane-1,5-diamine dihydrochloride is an endogenous diamine metabolite provided as research-grade reference material for analytical and synthetic laboratory use. It serves as an analytical standard in chromatographic and mass-spectrometry workflows and is offered in both solid and ready-to-use solution formats for flexibility in method development and QC applications.

  • Analytical standard suitable for HPLC, GC, and MS.
  • Available as a 10 mM solution in DMSO and in multiple solid pack sizes.
  • Molecular weight 175.10 g/mol; formula C5H16Cl2N2; CAS 1476-39-7.
  • Standard grade purity 98% for solid/reference material.
  • Documentation available: data sheet, COA, SDS, and spectral files.
  • Intended for research use only; follow handling and storage instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.