Learn More
Medchemexpress LLC Pentane-1,5-diamine dihydrochloride | 1476-39-7 | MFCD00012527 | 99.9% | 175.10 g/mol | C5H16Cl2N2 | 1 G
SDP
Catalog No. 5000301873
Encompass_Preferred
Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of pentane-1,5-diamine, offered as a high-purity solid reagent for research use. It serves as an analytical standard and reagent in biochemical research, metabolomics, and organic synthesis. Relevant identifiers include CAS 1476-39-7, molecular formula C5H16Cl2N2, and molecular weight 175.10 g/mol.
  • High purity: 99.9%.
  • Solid, white to off-white appearance.
  • Suitable as an analytical standard and reference material.
  • Applicable in biochemical, metabolomics, and synthetic chemistry workflows.
  • Available in multiple pack sizes for small- and large-scale use.
  • Store sealed at 4°C; follow solvent storage recommendations for solutions.
Catalog No. 50-003-01873 Supplier Medchemexpress LLC Supplier No. HYW0167501G
Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of pentane-1,5-diamine, offered as a high-purity solid reagent for research use. It serves as an analytical standard and reagent in biochemical research, metabolomics, and organic synthesis. Relevant identifiers include CAS 1476-39-7, molecular formula C5H16Cl2N2, and molecular weight 175.10 g/mol.
  • High purity: 99.9%.
  • Solid, white to off-white appearance.
  • Suitable as an analytical standard and reference material.
  • Applicable in biochemical, metabolomics, and synthetic chemistry workflows.
  • Available in multiple pack sizes for small- and large-scale use.
  • Store sealed at 4°C; follow solvent storage recommendations for solutions.