Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentane-1,5-diamine dihydrochloride | 1476-39-7 | MFCD00012527 | 99.9% | 175.10 g/mol | C5H16Cl2N2 | 100 MG
SDP

Catalog No. 5000303223
Encompass_Preferred

Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of 1,5-pentanediamine supplied as a solid analytical reagent. It serves as an endogenous metabolite reference and a building-block reagent for biochemical and synthetic applications.

  • White to off-white solid appearance.
  • Molecular weight 175.10 g/mol; formula C5H16Cl2N2.
  • High purity (reported 99.90%) suitable for analytical use.
  • Available in milligram-to-gram pack sizes and solution form.
  • SMILES: NCCCCCN.Cl.Cl for computational workflows.

Catalog No. 50-003-03223 Supplier Medchemexpress LLC Supplier No. HYW016750100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentane-1,5-diamine dihydrochloride is the dihydrochloride salt of 1,5-pentanediamine supplied as a solid analytical reagent. It serves as an endogenous metabolite reference and a building-block reagent for biochemical and synthetic applications.

  • White to off-white solid appearance.
  • Molecular weight 175.10 g/mol; formula C5H16Cl2N2.
  • High purity (reported 99.90%) suitable for analytical use.
  • Available in milligram-to-gram pack sizes and solution form.
  • SMILES: NCCCCCN.Cl.Cl for computational workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.