Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentane-1,5-diamine dihydrochloride | 1476-39-7 | MFCD00012527 | 99.9% | 175.10 g/mol | C5H16Cl2N2 | 50 MG
SDP

Catalog No. 5000300803
Encompass_Preferred

Pentane-1,5-diamine dihydrochloride is an endogenous metabolite provided as a high-purity research reagent for laboratory use. It is supplied as a white to off-white solid and is commonly used as a biochemical reagent or metabolite standard in research applications.

  • Purity 99.90%.
  • Molecular weight 175.10 g/mol.
  • Molecular formula C5H16Cl2N2.
  • Appearance white to off-white solid.
  • Soluble in DMSO; available as solid and as 10 mM solution in DMSO.

Catalog No. 50-003-00803 Supplier Medchemexpress LLC Supplier No. HYW01675050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pentane-1,5-diamine dihydrochloride is an endogenous metabolite provided as a high-purity research reagent for laboratory use. It is supplied as a white to off-white solid and is commonly used as a biochemical reagent or metabolite standard in research applications.

  • Purity 99.90%.
  • Molecular weight 175.10 g/mol.
  • Molecular formula C5H16Cl2N2.
  • Appearance white to off-white solid.
  • Soluble in DMSO; available as solid and as 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.