Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019002 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9002 10 mL
11-101-8166 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9002 Supplier MilliporeSigma Supelco Supplier No. 7687010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 500°F
CAS 109-66-0
Vapor Density 2.48 (vs. air)
Density 0.626 g/mL (at 25°C)
Flash Point -40°C
Physical Form Neat
Linear Formula CH3(CH2)3 CH3
MDL Number MFCD00009498
Melting Point -130°C
Boiling Point 35°C to 36°C
Type Analytical Standard
Quantity 10 mL
Formula Weight 72.15
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.