Learn More
Sigma Aldrich Fine Chemicals Biosciences Pentane anhydrous 991L
Catalog No. 502898338
Encompass
Pentane a linear aliphatic hydrocarbon is one of the main constituents of light straight-run (LSR) gasoline fraction in crude oils. Its isomerization can improve the octane number of gasoline. This has led to the development of a number of metal catalysts which can effectively isomerize n-pentane to iso-pentane.
Catalog No. 50-289-8338 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2367056X1L
Pentane a linear aliphatic hydrocarbon is one of the main constituents of light straight-run (LSR) gasoline fraction in crude oils. Its isomerization can improve the octane number of gasoline. This has led to the development of a number of metal catalysts which can effectively isomerize n-pentane to iso-pentane.