Learn More
Sigma Aldrich Fine Chemicals Biosciences Pentane puriss. p.a. 99.01L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 768711L
Encompass
Pentane (n-pentane) a hydrocarbon is a colorless liquid having 3 structural isomeric forms. It is a promising solvent for the disSOLUTIONon of oils fats and resins. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid. It isomerizes to iso-pentane in the presence of ZSM-5 (Zeolite Socony Mobil-5) zeolite catalysts under hydrogen and nitrogen atmosphere.
Catalog No. 50-289-8187