Learn More
Sigma Aldrich Fine Chemicals Biosciences Pentane suitable for HPLC
Catalog No. 502899379
Encompass
Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.
Catalog No. 50-289-9379 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 349562L
Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.