Learn More
Medchemexpress LLC Pentanediamide (ibrutinib-biotin conjugate) | 1599432-18-4 | 99.7% | 1097.37 | C56H80N12O9S | 1 ML
SDP
Catalog No. 5000214874
Encompass_Preferred
Ibrutinib-biotin is a biotinylated chemical probe that links ibrutinib to a long-chain biotin linker for use in biochemical and cell-based assays. It enables affinity-based target engagement studies and detection of Bruton's tyrosine kinase (BTK), and is supplied in solution and solid forms for flexible experimental workflows.
  • Biotinylated probe for pull-down and detection assays.
  • Reported BTK activity with IC50 0.755-1.02 nM.
  • Available as a 10 mM solution in DMSO and as solid quantities.
  • High purity as shown by certificate of analysis (>99.6%).
  • Soluble in DMSO (50 mg/mL) with recommended in vivo formulations provided.
  • Defined storage conditions for powder and in solvent to preserve stability.
Catalog No. 50-002-14874 Supplier Medchemexpress LLC Supplier No. HY10034210MM 1ML
Ibrutinib-biotin is a biotinylated chemical probe that links ibrutinib to a long-chain biotin linker for use in biochemical and cell-based assays. It enables affinity-based target engagement studies and detection of Bruton's tyrosine kinase (BTK), and is supplied in solution and solid forms for flexible experimental workflows.
  • Biotinylated probe for pull-down and detection assays.
  • Reported BTK activity with IC50 0.755-1.02 nM.
  • Available as a 10 mM solution in DMSO and as solid quantities.
  • High purity as shown by certificate of analysis (>99.6%).
  • Soluble in DMSO (50 mg/mL) with recommended in vivo formulations provided.
  • Defined storage conditions for powder and in solvent to preserve stability.