Learn More
Medchemexpress LLC Pentanediamide, N1-[4-[[4-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo... | 1599432-18-4 | MFCD30496724 | 99.7% | 1097.37 g/mol | C56H80N12O9S | 50 MG
SDP
Catalog No. 5000214967
Encompass_Preferred
Ibrutinib-biotin is a biotin-conjugated chemical probe derived from the BTK inhibitor ibrutinib. It features ibrutinib linked to biotin via a long-chain linker and is designed for target engagement, affinity pull-downs, and assay development directed at Bruton's tyrosine kinase (BTK).
  • Biotin-conjugated probe for affinity capture and pull-down assays.
  • Derived from a BTK inhibitor for target-specific engagement.
  • Low-nanomolar potency against BTK (IC50 0.755-1.02 nM).
  • High purity suitable for research applications (>99.6%).
  • Available as solid material and as a 10 mM solution in DMSO.
  • Molecular weight approximately 1,097.37 g/mol.
Catalog No. 50-002-14967 Supplier Medchemexpress LLC Supplier No. HY10034250MG
Ibrutinib-biotin is a biotin-conjugated chemical probe derived from the BTK inhibitor ibrutinib. It features ibrutinib linked to biotin via a long-chain linker and is designed for target engagement, affinity pull-downs, and assay development directed at Bruton's tyrosine kinase (BTK).
  • Biotin-conjugated probe for affinity capture and pull-down assays.
  • Derived from a BTK inhibitor for target-specific engagement.
  • Low-nanomolar potency against BTK (IC50 0.755-1.02 nM).
  • High purity suitable for research applications (>99.6%).
  • Available as solid material and as a 10 mM solution in DMSO.
  • Molecular weight approximately 1,097.37 g/mol.