Learn More
Medchemexpress LLC Pentanedioic-2,2,4,4-d4 acid | 19136-99-3 | 98.0% | 136.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW008820S150MG
Encompass_Preferred
Glutaric acid-d4 is the deuterium labeled Glutaric acid. Glutaric acid, a C5 dicarboxylic acid, acts as an intermediate in the catabolic pathways of lysine and tryptophan. It influences pericyte contractility and migration, and serves as an indicator of glutaric aciduria type I.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52132