Learn More
Medchemexpress LLC Pentanedioic acid, 1-[2-[[[8-[(2,6-dimethylphenyl)methyl]amino]...ethyl] ester | 1228559-81-6 | 99.7% | 480.56 g/mol | C26H32N4O5 | 5 MG
SDP
Catalog No. 5000221581
Encompass_Preferred
Linaprazan glurate is an orally active ethyl-ester prodrug of linaprazan that acts as a potassium-competitive acid blocker. It is supplied as a white to off-white solid for research and pharmacological studies on gastric acid secretion, ADME, and therapeutic evaluation.
  • Potent potassium-competitive acid blocker.
  • Orally active prodrug of linaprazan.
  • High purity (~99.7%).
  • White to off-white solid suitable for analytical and in vivo research.
  • Available in small packaged amounts for screening and dose-finding studies.
  • Molecular formula C26H32N4O5; molecular weight 480.56 g/mol.
Catalog No. 50-002-21581 Supplier Medchemexpress LLC Supplier No. HY1455785MG
Linaprazan glurate is an orally active ethyl-ester prodrug of linaprazan that acts as a potassium-competitive acid blocker. It is supplied as a white to off-white solid for research and pharmacological studies on gastric acid secretion, ADME, and therapeutic evaluation.
  • Potent potassium-competitive acid blocker.
  • Orally active prodrug of linaprazan.
  • High purity (~99.7%).
  • White to off-white solid suitable for analytical and in vivo research.
  • Available in small packaged amounts for screening and dose-finding studies.
  • Molecular formula C26H32N4O5; molecular weight 480.56 g/mol.