Learn More
Medchemexpress LLC Pentanedioic acid, 1,5-bis(2,5-dioxo-1-pyrrolidinyl) ester | 79642-50-5 | ≥98.0% | 326.26 | 25 MG
SDP
Catalog No. 5000438165
Encompass_Preferred
DSG Crosslinker is an amine-reactive and noncleavable protein crosslinker. It enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive and noncleavable protein crosslinker.
  • Enables stable protein-protein, peptide, and biomolecule conjugation.
Catalog No. 50-004-38165 Supplier Medchemexpress LLC Supplier No. 25MGHY114697
DSG Crosslinker is an amine-reactive and noncleavable protein crosslinker. It enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive and noncleavable protein crosslinker.
  • Enables stable protein-protein, peptide, and biomolecule conjugation.