Learn More
Medchemexpress LLC Pentanedioic acid, 2-oxo-, potassium salt (1:1) | 997-43-3 | 98.0% | 185.20 | 1 G
SDP
Catalog No. 5000454519
Encompass_Preferred
2-Ketoglutaric acid potassium is an intermediate in ATP/GTP production within the Krebs cycle and a reversible, orally active inhibitor of tyrosinase (IC50 = 15 mM). It acts as the primary carbon skeleton for nitrogen-assimilatory reactions. It suppresses abnormal intestinal permeability, prevents tight junction protein delocalization, and inhibits TNFα expression in vitro and in vivo. It binds to TAK1, hindering TRAF6-TAK1 interaction, and alleviates inflammatory bowel disease (IBD) symptoms and gut microbiota dysbiosis.
  • Reduces and detoxifies ammonia levels.
  • Protective against lipid peroxidation.
  • Neuroprotective effect against cyanide poisoning.
  • Acts as a precursor for amino acids and nucleotides.
  • Improves gut microbiota dysbiosis.
Catalog No. 50-004-54519 Supplier Medchemexpress LLC Supplier No. HYW013636C1G
2-Ketoglutaric acid potassium is an intermediate in ATP/GTP production within the Krebs cycle and a reversible, orally active inhibitor of tyrosinase (IC50 = 15 mM). It acts as the primary carbon skeleton for nitrogen-assimilatory reactions. It suppresses abnormal intestinal permeability, prevents tight junction protein delocalization, and inhibits TNFα expression in vitro and in vivo. It binds to TAK1, hindering TRAF6-TAK1 interaction, and alleviates inflammatory bowel disease (IBD) symptoms and gut microbiota dysbiosis.
  • Reduces and detoxifies ammonia levels.
  • Protective against lipid peroxidation.
  • Neuroprotective effect against cyanide poisoning.
  • Acts as a precursor for amino acids and nucleotides.
  • Improves gut microbiota dysbiosis.