Learn More
Medchemexpress LLC Pentanedioic acid, 3-oxo-, 1,5-diethyl ester | 105-50-0 | MFCD00009200 | ≥97.0% | 202.21 g/mol | C9H14O5 | 1 KG
SDP
Catalog No. 5000303894
Encompass_Preferred
Diethyl 3-oxopentanedioate is an organic biochemical reagent used as a building block in organic synthesis and as a reagent for biochemical research. It is supplied for research use only and is characterized by high purity, a liquid appearance, and available documentation including a datasheet and safety data sheet.
  • Has purity ≥97.0%.
  • Molecular formula C9H14O5 and molecular weight 202.21 g/mol.
  • Liquid appearance, colorless to light yellow, density 1.113 g/cm3.
  • Available in multiple pack sizes up to 1 kg.
  • Supplied with downloadable datasheet and safety data sheet.
  • Suitable as a building block in organic synthesis and for biochemical assays.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-03894 Supplier Medchemexpress LLC Supplier No. HYW0147991KG
Diethyl 3-oxopentanedioate is an organic biochemical reagent used as a building block in organic synthesis and as a reagent for biochemical research. It is supplied for research use only and is characterized by high purity, a liquid appearance, and available documentation including a datasheet and safety data sheet.
  • Has purity ≥97.0%.
  • Molecular formula C9H14O5 and molecular weight 202.21 g/mol.
  • Liquid appearance, colorless to light yellow, density 1.113 g/cm3.
  • Available in multiple pack sizes up to 1 kg.
  • Supplied with downloadable datasheet and safety data sheet.
  • Suitable as a building block in organic synthesis and for biochemical assays.
  • For research use only; not for human or clinical use.