Learn More
Medchemexpress LLC Pentanedioic acid, 3-oxo-, 1,5-diethyl ester | 105-50-0 | MFCD00009200 | ≥97.0% | 202.21 g/mol | C9H14O5 | 500 G
SDP
Catalog No. 5000303895
Encompass_Preferred
Diethyl 3-oxopentanedioate is a diethyl ester reagent used in organic synthesis and biochemical research. Supplied as a colorless to light yellow liquid with typical purity ≥97.0%, it serves as a synthetic building block and assay reagent for life-science applications.
  • Used as a biochemical reagent and synthetic building block.
  • Typical purity ≥97.0%.
  • Liquid appearance; density 1.113 g/cm3.
  • Molecular weight 202.21 g/mol; formula C9H14O5.
  • Available in multiple pack sizes including 500 g.
Catalog No. 50-003-03895 Supplier Medchemexpress LLC Supplier No. HYW014799500G
Diethyl 3-oxopentanedioate is a diethyl ester reagent used in organic synthesis and biochemical research. Supplied as a colorless to light yellow liquid with typical purity ≥97.0%, it serves as a synthetic building block and assay reagent for life-science applications.
  • Used as a biochemical reagent and synthetic building block.
  • Typical purity ≥97.0%.
  • Liquid appearance; density 1.113 g/cm3.
  • Molecular weight 202.21 g/mol; formula C9H14O5.
  • Available in multiple pack sizes including 500 g.