Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentanedioic acid, 3-oxo-, 1,5-dimethyl ester | 1830-54-2 | MFCD00008462 | 97.0% | 174.15 g·mol⁻¹ | C7H10O5 | 1 KG
SDP

Catalog No. 5000301227
Encompass_Preferred

Dimethyl acetone-1,3-dicarboxylate (dimethyl-3-oxoglutarate) is an organic biochemical reagent used as a small-molecule building block in synthetic chemistry and biochemical assays. Supplied as a purified dimethyl ester, it is intended for research use in life-science applications.

  • High purity (97.0%).
  • Molecular formula C7H10O5.
  • Molecular weight 174.15 g·mol⁻¹.
  • Available in multiple pack sizes for flexible ordering.
  • Includes datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-003-01227 Supplier Medchemexpress LLC Supplier No. HYW0168011KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl acetone-1,3-dicarboxylate (dimethyl-3-oxoglutarate) is an organic biochemical reagent used as a small-molecule building block in synthetic chemistry and biochemical assays. Supplied as a purified dimethyl ester, it is intended for research use in life-science applications.

  • High purity (97.0%).
  • Molecular formula C7H10O5.
  • Molecular weight 174.15 g·mol⁻¹.
  • Available in multiple pack sizes for flexible ordering.
  • Includes datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.