Learn More
Medchemexpress LLC Pentanehydrazide (Valeric acid hydrazide) | 38291-82-6 | ≥95.0% | 116.16 | 100 G
SDP
Catalog No. 5000455050
Encompass_Preferred
Pentanehydrazide, also known as Valeric acid hydrazide, is a biochemical reagent. It functions as an effective alkylating agent in the copper-catalyzed oxidative cyclization of N-arylacrylamides and participates in tandem acylation reactions.
  • Acts as an effective alkylating agent.
  • Involved in copper-catalyzed oxidative cyclization reactions.
  • Participates in tandem acylation reactions.
  • Suitable for research use only.
Catalog No. 50-004-55050 Supplier Medchemexpress LLC Supplier No. HYW012890100G
Pentanehydrazide, also known as Valeric acid hydrazide, is a biochemical reagent. It functions as an effective alkylating agent in the copper-catalyzed oxidative cyclization of N-arylacrylamides and participates in tandem acylation reactions.
  • Acts as an effective alkylating agent.
  • Involved in copper-catalyzed oxidative cyclization reactions.
  • Participates in tandem acylation reactions.
  • Suitable for research use only.