Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentanoic acid, 4,4-dimethyl-3-oxo-, ethyl ester | 17094-34-7 | MFCD00042886 | 98.3% | 172.22 g/mol | C9H16O3 | 25 G
SDP

Catalog No. 5000209018
Encompass_Preferred

Ethyl pivaloylacetate is a β-ketoester reagent used as a substrate to evaluate ketoreductase activity and stereoselectivity. Consult the SDS and COA for safety, purity, and handling details.

  • β-ketoester reagent for enzymatic assays and stereoselectivity studies
  • high purity: 98.3%
  • molecular formula C9H16O3; molecular weight 172.22 g/mol
  • available in research pack sizes, including 25 G
  • recommended storage: pure form -20°C (3 years) or 4°C (2 years)
  • safety and handling information available in SDS and COA

Catalog No. 50-002-09018 Supplier Medchemexpress LLC Supplier No. 25GHYW076971
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl pivaloylacetate is a β-ketoester reagent used as a substrate to evaluate ketoreductase activity and stereoselectivity. Consult the SDS and COA for safety, purity, and handling details.

  • β-ketoester reagent for enzymatic assays and stereoselectivity studies
  • high purity: 98.3%
  • molecular formula C9H16O3; molecular weight 172.22 g/mol
  • available in research pack sizes, including 25 G
  • recommended storage: pure form -20°C (3 years) or 4°C (2 years)
  • safety and handling information available in SDS and COA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.