Learn More
Medchemexpress LLC Pentanoic acid, 5-(2,5-dimethylphenoxy)-2,2-di(methyl-d3)- | 1184986-45-5 | 98.7% | 1 MG
SDP
Catalog No. 5000406930
Encompass_Preferred
Gemfibrozil-d6 is the deuterium labeled form of Gemfibrozil, a known activator of PPAR-α and a lipid-lowering agent. It also functions as a nonselective inhibitor of several P450 isoforms. This compound is intended for research use only and serves as a valuable tool in scientific investigations, particularly as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium labeled form of gemfibrozil
  • Activator of PPAR-α
  • Used as a lipid-lowering agent
  • Nonselective inhibitor of several P450 isoforms
  • Suitable as a tracer in research
  • Ideal as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes for pharmacokinetic studies
Catalog No. 50-004-06930 Supplier Medchemexpress LLC Supplier No. HYB0258S1MG
Gemfibrozil-d6 is the deuterium labeled form of Gemfibrozil, a known activator of PPAR-α and a lipid-lowering agent. It also functions as a nonselective inhibitor of several P450 isoforms. This compound is intended for research use only and serves as a valuable tool in scientific investigations, particularly as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium labeled form of gemfibrozil
  • Activator of PPAR-α
  • Used as a lipid-lowering agent
  • Nonselective inhibitor of several P450 isoforms
  • Suitable as a tracer in research
  • Ideal as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes for pharmacokinetic studies