Learn More
Medchemexpress LLC Pentanoic acid, 5-bromo- | 2067-33-6 | MFCD00004414 | 97.0% | 181.03 g·mol⁻¹ | C5H9BrO2 | 50 G
SDP
Catalog No. 5000301112
Encompass_Preferred
5-Bromopentanoic acid is a bromo-substituted pentanoic acid used as a chemical intermediate in organic synthesis, including preparation of inhibitors active against aminoglycoside-resistant bacteria and other small-molecule derivatives. It is supplied as a solid suitable for research and scale-up.
  • Bromo-substituted pentanoic acid useful as a synthetic intermediate.
  • Molecular formula C5H9BrO2; molecular weight 181.03 g·mol⁻¹.
  • Solid physical state; melting point ≈ 38-40 °C.
  • Available in common laboratory pack sizes (50 g, 100 g, 500 g).
  • Typical purity 97.0% by NMR as reported on batch COA.
  • Suitable for small-molecule synthesis and medicinal chemistry research.
Catalog No. 50-003-01112 Supplier Medchemexpress LLC Supplier No. HYW01645650G
5-Bromopentanoic acid is a bromo-substituted pentanoic acid used as a chemical intermediate in organic synthesis, including preparation of inhibitors active against aminoglycoside-resistant bacteria and other small-molecule derivatives. It is supplied as a solid suitable for research and scale-up.
  • Bromo-substituted pentanoic acid useful as a synthetic intermediate.
  • Molecular formula C5H9BrO2; molecular weight 181.03 g·mol⁻¹.
  • Solid physical state; melting point ≈ 38-40 °C.
  • Available in common laboratory pack sizes (50 g, 100 g, 500 g).
  • Typical purity 97.0% by NMR as reported on batch COA.
  • Suitable for small-molecule synthesis and medicinal chemistry research.