Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pentanoic acid, 5-(dimethylamino)-, (10Z,13Z)-1-(9Z,12Z)-9,12-octadecadien-1-yl ester | 1226909-66-5 | 98.0% | 656.12 | C44H81NO2 | 10 MG
SDP

Catalog No. 5000287594
Encompass_Preferred

DLin-M-C4-DMA is a cationic ionizable lipid used to formulate lipid nanoparticles for delivery of nucleic acids, including siRNA and mRNA. It is supplied for research use and is characterized by a long aliphatic structure that enables incorporation into lipid-based delivery systems. Handle and store under recommended cold-chain conditions to preserve integrity.

  • Cationic lipid for delivery of nucleic acids (siRNA, mRNA).
  • Suitable for formulation of lipid nanoparticles and transfection reagents.
  • Molecular formula C44H81NO2 and molecular weight 656.12.
  • Purity (NMR) 98.0%.
  • Available in small mg quantities and as 10 mM solutions in DMSO.
  • Storage: pure form -20°C (long term) and 4°C (short term); solutions -80°C recommended.
  • Intended for research use only; not for clinical or therapeutic use.

Catalog No. 50-002-87594 Supplier Medchemexpress LLC Supplier No. HY15480410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-M-C4-DMA is a cationic ionizable lipid used to formulate lipid nanoparticles for delivery of nucleic acids, including siRNA and mRNA. It is supplied for research use and is characterized by a long aliphatic structure that enables incorporation into lipid-based delivery systems. Handle and store under recommended cold-chain conditions to preserve integrity.

  • Cationic lipid for delivery of nucleic acids (siRNA, mRNA).
  • Suitable for formulation of lipid nanoparticles and transfection reagents.
  • Molecular formula C44H81NO2 and molecular weight 656.12.
  • Purity (NMR) 98.0%.
  • Available in small mg quantities and as 10 mM solutions in DMSO.
  • Storage: pure form -20°C (long term) and 4°C (short term); solutions -80°C recommended.
  • Intended for research use only; not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.