Learn More
Bachem H-Phe-Phe-Phe-Phe-Phe-OH
SDP
Supplier:  Bachem 4012151.0100
Encompass_Preferred
H-Phe-Phe-Phe-Phe-Phe-OH, 65757-10-0, C45H47N5O6, Mr 753.90, 100mg. Pentaphenylalanine and tetraphenylalanine could be assembled into ordered autofluorescent elongated structures.
Catalog No. 50-194-7519