Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaphenylcyclopentadiene, 99%

Catalog No. AAH2722303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2722303 1 g
AAH2722306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2722303 Supplier Thermo Scientific Chemicals Supplier No. H2722303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2519-10-0 | C35H26 | 446.593 g/mol

1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

Solubility
Soluble in pyridine 5 mg/ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519-10-0
Molecular Formula C35H26
Molecular Weight (g/mol) 446.593
MDL Number MFCD09909572
InChI Key YGLVWOUNCXBPJF-UHFFFAOYSA-N
PubChem CID 635023
IUPAC Name (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene
SMILES C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 254°C to 257°C
Quantity 1 g
Synonym 1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene, pentaphenylcyclopentadiene, cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, 2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl benzene, acmc-1cek1, 2,3,4,5-tetraphenyl-1-cyclopenta-1,4-dienyl benzene, benzene, 1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dienyl benzene
Solubility Information Soluble in pyridine 5mg/ml.
Formula Weight 446.59
Percent Purity 99%
Chemical Name or Material Pentaphenylcyclopentadiene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.