Learn More
AdipoGen Penthiopyrad,Chemical. CAS: 183675-82-3
SDP
Supplier:  AdipoGen CDXP0166M025
Encompass_Preferred
Penthiopyrad, AdipoGen Life Sciences. Chemical. CAS: 183675-82-3. Formula: C16H20F3N3OS. MW: 359.4. Synthetic. Systemic broad spectrum fungicide. Interferes with fungal respiration by inhibition of Complex II: succinate-dehydrogenase (SDH). Compound can be used as analytical reference material. 183675-82-3
Catalog No. 50-117-7686