Learn More
Medchemexpress LLC HY-100143 5mg Medchemexpress, Pentiapine CAS:81382-51-6 Purity:>98%
SDP
Catalog No. 501931731
Encompass_Preferred
Medchemexpress, HY-100143 5mg Pentiapine CAS:81382-51-6 Pentiapine is a novel dopamine release inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1731 Supplier Medchemexpress LLC Supplier No. HY1001435MG
Medchemexpress, HY-100143 5mg Pentiapine CAS:81382-51-6 Pentiapine is a novel dopamine release inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.