Learn More
Medchemexpress LLC Pentifylline (1-hexyltheobromine) | 1028-33-7 | MFCD00041424 | 99.2% | 264.32 g/mol | C13H20N4O2 | 250 MG
SDP
Catalog No. 5000282310
Encompass_Preferred
Pentifylline (1-Hexyltheobromine) is a vasodilator research compound with anti-inflammatory properties commonly used in laboratory and analytical applications. It modulates platelet function and vascular biology, and is provided as a high-purity powder with specified storage conditions for research use.
  • Vasodilator with anti-inflammatory activity.
  • Inhibits platelet aggregation and thromboxane A2 synthesis.
  • Reduces release of free radicals and oxidative stress.
  • Decreases type I collagen and laminin production in fibroblasts.
  • Supplied as a high-purity solid suitable for analytical and research use.
  • Defined storage conditions for powder and solution stability.
Catalog No. 50-002-82310 Supplier Medchemexpress LLC Supplier No. HYW273690250MG
Pentifylline (1-Hexyltheobromine) is a vasodilator research compound with anti-inflammatory properties commonly used in laboratory and analytical applications. It modulates platelet function and vascular biology, and is provided as a high-purity powder with specified storage conditions for research use.
  • Vasodilator with anti-inflammatory activity.
  • Inhibits platelet aggregation and thromboxane A2 synthesis.
  • Reduces release of free radicals and oxidative stress.
  • Decreases type I collagen and laminin production in fibroblasts.
  • Supplied as a high-purity solid suitable for analytical and research use.
  • Defined storage conditions for powder and solution stability.