Learn More
Medchemexpress LLC Pentigetide | 62087-72-3 | 99.4% | 588.57 | 5 MG
SDP
Catalog No. 5000397284
Encompass_Preferred
Pentigetide is a synthetic peptide (Asp-Ser-Asp-Pro-Arg or D-S-D-P-R), also known as Human IgE Pentapeptide. This peptide cannot produce IgE inhibition in several different systems because the aspartic acids in the original peptide have been changed into an asparagine.
  • Synthetic peptide
  • Also known as human IgE pentapeptide
  • Does not produce IgE inhibition due to altered aspartic acids
Catalog No. 50-003-97284 Supplier Medchemexpress LLC Supplier No. HYP37465MG
Pentigetide is a synthetic peptide (Asp-Ser-Asp-Pro-Arg or D-S-D-P-R), also known as Human IgE Pentapeptide. This peptide cannot produce IgE inhibition in several different systems because the aspartic acids in the original peptide have been changed into an asparagine.
  • Synthetic peptide
  • Also known as human IgE pentapeptide
  • Does not produce IgE inhibition due to altered aspartic acids