Learn More
Cayman Chemical Pentanoic Acid-d9, Purity: ≥98%, CAS: 115871-50-6, 250 mg
SDP
Catalog No. 502941867
Encompass_Preferred
An internal standard for the quantification of pentanoic acid by GC- or LC-MS
Catalog No. 50-294-1867 Supplier Cayman Chemical Supplier No. 28870250 MG
An internal standard for the quantification of pentanoic acid by GC- or LC-MS