Learn More
Cayman Chemical Pentanoic Acid-d9, Purity: ≥98%, CAS: 115871-50-6, 50 mg
SDP
Catalog No. 502937030
Encompass_Preferred
An internal standard for the quantification of pentanoic acid by GC- or LC-MS
Catalog No. 50-293-7030 Supplier Cayman Chemical Supplier No. 2887050 MG
An internal standard for the quantification of pentanoic acid by GC- or LC-MS