Learn More
Cayman Chemical Pentnoyl-Coenzyme AsodIum 1mg
SDP
Catalog No. 502942926
Encompass_Preferred
A saturated acyl CoA and metabolite of 3-oxovalproyl-CoA; hepatic levels of pentanoyl-CoA and propionyl-CoA are increased in an Apc+/Min-FCCC mouse model of spontaneous colorectal cancer fed a low- or high-fat diet
Catalog No. 50-294-2926 Supplier Cayman Chemical Supplier No. 404601 MG
A saturated acyl CoA and metabolite of 3-oxovalproyl-CoA; hepatic levels of pentanoyl-CoA and propionyl-CoA are increased in an Apc+/Min-FCCC mouse model of spontaneous colorectal cancer fed a low- or high-fat diet