Learn More
Cayman Chemical Pentanoyl-Coenzyme A (sodium salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502943242
Encompass_Preferred
A saturated acyl CoA and metabolite of 3-oxovalproyl-CoA; hepatic levels of pentanoyl-CoA and propionyl-CoA are increased in a model of spontaneous colorectal cancer using Apc+/Min-FCCC mice fed a low- or high-fat diet
Catalog No. 50-294-3242 Supplier Cayman Chemical Supplier No. 404605 MG
A saturated acyl CoA and metabolite of 3-oxovalproyl-CoA; hepatic levels of pentanoyl-CoA and propionyl-CoA are increased in a model of spontaneous colorectal cancer using Apc+/Min-FCCC mice fed a low- or high-fat diet